missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sorbitol, Powder, FCC, 91-100.5%, Spectrum™ Chemical
S1523, 50-70-4, C6H14O6
$958.52 - $6,244.52
Chemical Identifiers
| CAS | 50-70-4 |
|---|---|
| Molecular Formula | C6H14O6 |
| Molecular Weight (g/mol) | 182.17 |
| InChI Key | FBPFZTCFMRRESA-UHFFFAOYNA-N |
| IUPAC Name | hexane-1,2,3,4,5,6-hexol |
| SMILES | OCC(O)C(O)C(O)C(O)CO |