missing translation for 'onlineSavingsMsg'
Learn More

Sorbitol, Powder, FCC, 91-100.5%, Spectrum™ Chemical

Catalog No. 18606957
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
45 kg
Packaging:
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606957 2.5 kg Poly Pail
18606958 12 kg Poly Pail
18606959 45 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18606957 Supplier Spectrum Chemical Mfg Cor Supplier No. S15232.5KG
Get Up to $500 in Chemicals with Purchase
View all promotions

S1523, 50-70-4, C6H14O6

Spectrum™ Chemical Sorbitol, Powder, FCC is used is as a sugar substitute sweetener in many foods. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 50-70-4
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
IUPAC Name hexane-1,2,3,4,5,6-hexol
SMILES OCC(O)C(O)C(O)C(O)CO

Specifications

CAS Max % 1
pH 3.5 to 7.0
Boiling Point 295°C (563°F)
Ignition Residue 0.001
Packaging Poly Pail
Quantity 2.5 kg
Percent Purity 91 to 100.5%
Grade FCC
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Sorbitol, Powder