missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sorbitan Tristearate, Spectrum™ Chemical
Spécifications
Spécifications
| CAS Max % | 1 |
| Packaging | Poly Bottle |
| Quantity | 2.5 kg |
| Saponification Value | 176 - 188 |
| Grade | Reagent |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.