missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sorbitan Trioleate, Spectrum™ Chemical
Description
Spectrum™ Chemical Sorbitan Trioleate
Specifications
Specifications
| CAS Max % | 1 |
| Packaging | Amber Glass Bottle |
| Quantity | 200 L |
| Viscosity | Approx. 210 cps |
| Grade | Ungraded |
| Physical Form | Viscous Liquid or Low Melting Solid |
| Chemical Name or Material | Sorbitan Trioleate |