missing translation for 'onlineSavingsMsg'
Learn More

Sorbitan Monostearate, FCC, 27-34%, Spectrum™ Chemical

Catalog No. 18606932
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606932 500 g Poly Bottle
18606933 2.5 kg Poly Bottle
18606934 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18606932 Supplier Spectrum Chemical Mfg Cor Supplier No. S1318500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

S1318, 1338-41-6, C24H46O6

Spectrum™ Chemical Sorbitan Monostearate, FCC is used as an emulsifier to keep oils and water mixed. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1338-41-6
Molecular Formula C24H46O6
Molecular Weight (g/mol) 430.63
MDL Number MFCD00005366,MFCD00005366
InChI Key HVUMOYIDDBPOLL-IIZJTUPISA-N
IUPAC Name 2-[(2R,3S,4R)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 500 g
Saponification Value 147-157
Percent Purity 27 to 34%
Grade FCC
Physical Form Powder or Waxy Solid
Chemical Name or Material Sorbitan Monostearate