missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sorbitan Monooleate, NF, 72-78%, Spectrum™ Chemical
SPA83, 1338-43-8, C24H44O6
$1,418.15 - $22,681.04
Chemical Identifiers
| CAS | 1338-43-8 |
|---|---|
| Molecular Formula | C24H44O6 |
| Molecular Weight (g/mol) | 428.61 |
| MDL Number | MFCD00080948 |
| InChI Key | NWGKJDSIEKMTRX-NZAQQJATSA-N |
| IUPAC Name | 2-[(2R,3S,4R)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl (9Z)-octadec-9-enoate |
| SMILES | CCCCCCCC\C=C/CCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
18606924
|
N/A | 4 L | Amber Glass Bottle |
Each for $1,418.15
|
|
||||
|
18606925
|
N/A | 20 L | Amber Glass Bottle |
Each for $4,326.21
|
|
||||
|
18606926
|
N/A | 200 L | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Sorbitan Monooleate, NF is used as an emulsifier and nonionic surfactant in pharmaceutical formulations. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Chemical Identifiers
| 1338-43-8 | |
| 428.61 | |
| NWGKJDSIEKMTRX-NZAQQJATSA-N | |
| CCCCCCCC\C=C/CCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O |
| C24H44O6 | |
| MFCD00080948 | |
| 2-[(2R,3S,4R)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl (9Z)-octadec-9-enoate |
Specifications
| 1338-43-8 | |
| 0.005 | |
| C24H44O6 | |
| MFCD00080948 | |
| CCCCCCCC\C=C/CCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O | |
| 428.61 | |
| NF |
| 1 | |
| 145-160 | |
| 0.01 | |
| NWGKJDSIEKMTRX-NZAQQJATSA-N | |
| 2-[(2R,3S,4R)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl (9Z)-octadec-9-enoate | |
| 72 to 78% | |
| Sorbitan Monooleate |