missing translation for 'onlineSavingsMsg'
Learn More

Sorbitan Monooleate, NF, 72-78%, Spectrum™ Chemical

Catalog No. 18606926
Change view
Click to view available options
Quantity:
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606926 200 L Amber Glass Bottle
18606924 4 L Amber Glass Bottle
18606925 20 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18606926 Supplier Spectrum Chemical Mfg Cor Supplier No. SPA83200LTBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SPA83, 1338-43-8, C24H44O6

Spectrum™ Chemical Sorbitan Monooleate, NF is used as an emulsifier and nonionic surfactant in pharmaceutical formulations. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1338-43-8
Molecular Formula C24H44O6
Molecular Weight (g/mol) 428.61
MDL Number MFCD00080948
InChI Key NWGKJDSIEKMTRX-NZAQQJATSA-N
IUPAC Name 2-[(2R,3S,4R)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl (9Z)-octadec-9-enoate
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O

Specifications

CAS Max % 1
Quantity 200 L
Ignition Residue 0.005
Packaging Amber Glass Bottle
Saponification Value 145-160
Water 0.01
Percent Purity 72 to 78%
Grade NF
Chemical Name or Material Sorbitan Monooleate