missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sorbitan Monooleate, NF, 72-78%, Spectrum™ Chemical
SPA83, 1338-43-8, C24H44O6
Supplier: Spectrum Chemical Mfg Cor SPA8320LT
| Quantity | 20 L |
|---|---|
| Packaging | Amber Glass Bottle |
Description
Spectrum™ Chemical Sorbitan Monooleate, NF is used as an emulsifier and nonionic surfactant in pharmaceutical formulations. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Chemical Identifiers
| 1338-43-8 | |
| 428.61 | |
| NWGKJDSIEKMTRX-NZAQQJATSA-N | |
| CCCCCCCC\C=C/CCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O |
| C24H44O6 | |
| MFCD00080948 | |
| 2-[(2R,3S,4R)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl (9Z)-octadec-9-enoate |
Specifications
| Sorbitan Monooleate | |
| 1 | |
| 0.005 | |
| 145-160 | |
| 0.01 | |
| NWGKJDSIEKMTRX-NZAQQJATSA-N | |
| 2-[(2R,3S,4R)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl (9Z)-octadec-9-enoate | |
| 72 to 78% |
| 1338-43-8 | |
| 20 L | |
| Amber Glass Bottle | |
| C24H44O6 | |
| MFCD00080948 | |
| CCCCCCCC\C=C/CCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O | |
| 428.61 | |
| NF |