missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sorbic Acid Standard, 300 ppm C6H8O2, Ricca Chemical
Supplier: Ricca Chemical Company R7986300100A
Specifications
| Sorbic Acid Standard, 300ppm | |
| 110-44-1 , 7732-18-5 | |
| 100.0,0.03 | |
| C6H8O2 | |
| Colorless | |
| 100°C | |
| Miscible with water | |
| Standard | |
| C\C=C\C=C\C(O)=O | |
| 112.13 | |
| 643460 | |
| Laboratory |
| Natural Poly Bottle | |
| 98.0,0.03 | |
| Liquid | |
| MFCD00002703 | |
| 0.0°C | |
| GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation | |
| Calibration Standard | |
| WSWCOQWTEOXDQX-MQQKCMAXSA-N | |
| (2E,4E)-hexa-2,4-dienoic acid | |
| 100 mL | |
| CHEBI:38358 |
Chemical Identifiers
| 110-44-1 | |
| 112.13 | |
| WSWCOQWTEOXDQX-MQQKCMAXSA-N | |
| CHEBI:38358 | |
| C\C=C\C=C\C(O)=O |
| C6H8O2 | |
| MFCD00002703 | |
| 643460 | |
| (2E,4E)-hexa-2,4-dienoic acid |