missing translation for 'onlineSavingsMsg'
Learn More

Sorbic Acid Standard, 100 ppm C6H8O2, Ricca Chemical

Supplier:  Ricca Chemical Company R7986100100A

Catalog No. R7986100100


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sorbic Acid Standard, 100ppm, Ricca Chemical
110-44-1 , 7732-18-5
100.0,0.01
C6H8O2
Colorless
100°C
Miscible with water
Standard
C\C=C\C=C\C(O)=O
112.13
643460
Laboratory
Natural Poly Bottle
98.0,0.01
Liquid
MFCD00002703
0.0°C
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Calibration Standard
WSWCOQWTEOXDQX-MQQKCMAXSA-N
(2E,4E)-hexa-2,4-dienoic acid
100 mL
CHEBI:38358

Chemical Identifiers

110-44-1
112.13
WSWCOQWTEOXDQX-MQQKCMAXSA-N
CHEBI:38358
C\C=C\C=C\C(O)=O
C6H8O2
MFCD00002703
643460
(2E,4E)-hexa-2,4-dienoic acid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions