missing translation for 'onlineSavingsMsg'
Learn More

Sorbent, Bulk Silica, 10 grams, Solstice

Catalog No. 6002994
Change view
Click to view available options
Quantity:
1 Ea.
Packaging:
Fiberboard Box
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6002994 1 Ea. Fiberboard Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6002994 Supplier Solstice Supplier No. 9954

Bulk Silica, 10 grams

  • Wide selection of support materials and multiple cartridge/column sizes
  • Every stationary phase batch tested to ensure lot-to-lot reproducibility
  • Reliable quality

Chemical Identifiers

CAS 71889-02-6
Molecular Formula C18H37Cl3O2Si2
Molecular Weight (g/mol) 448.012
InChI Key VYTRPUAMVCWPLO-UHFFFAOYSA-N
PubChem CID 172932
IUPAC Name dioxosilane;trichloro(octadecyl)silane
SMILES CCCCCCCCCCCCCCCCCC[Si](Cl)(Cl)Cl.O=[Si]=O

Specifications

Quantity 1 Ea.
Packaging Fiberboard Box
Grade Analytical
Chemical Name or Material Sorbent, Bulk Silica, 10 grams