missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sonicated DNAs for Hybridization Studies

Catalog No. 45001363
Change view
Click to view available options
Product Type:
Sonicated Calf Thymus DNA
Sonicated Salmon Sperm DNA
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
45001363 Sonicated Salmon Sperm DNA
45001362 Sonicated Calf Thymus DNA
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45001363 Supplier Cytiva Supplier No. 27456501
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Suitable for blocking non-specific binding sites in hybridization experiments to reduce background. Sonicated DNAs for Hybridization Studies are purified by phenol extraction and freeze dried.

  • Fragmented DNA with a size range from 200-6000 nucleotides
  • Purified by phenol extraction and freeze dried
  • Tested for absence of nuclease
  • DNA isolated from Calf thymus is sonicated to fragment the DNA to an average size of approximately 3000 bp

Specifications

Format Solid
Product Type Sonicated Salmon Sperm DNA
Content And Storage -20°C
Quantity 100/Pk.
For Use With (Application) Hybridisation experiments