Learn More
SOMNI Scientific™ SOMNI Scientific™ SOMNI Adu Coaxial Circuit
Numéro de catalogue. 04777204
Change view
Click to view available options
Length (English):
60 in.
Numéro de catalogue. Length (English)
04777204 60 in.
1 options
Numéro de catalogue. 04777204 Fournisseur SOMNI Scientific™ Code fournisseur BC13000
Il en reste null
The tube within a tube design eliminates twisting of the inspiratory and expiratory limbs and provides heat and moisture conservation. PVC, Phthalate and Latex Free
  • A tube within a tube design that eliminates twisting of the inspiratory and expiratory limbs.
  • PVC, Phthalate and Latex Free
  • Provides heat and moisture conservation
Spécifications
For Use With (Application) Research Animal Inhalant Anesthesia
Length (English) 60 in.