missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Solvent-Vented™ and Safety-Labeled Wash Bottles
GREENER_CHOICE

Pre-printed with Chemical name and formula. Fisherbrand™ Solvent-Vented™ and Safety-Labeled Wash Bottles have a patented valve which eliminates drips and allows bottle to dispense upright or inverted.

Supplier:  Fisherbrand™ FB0340934

 View more versions of this product

Catalog No. FB0340934



Only null left

Description

Description

  • Wide mouth 2 in. (53mm) bottle for easy refilling
  • Polypropylene closure with leakproof design, color-coded to aid in easy identification
  • Translucent, LDPE solvent-resistant; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
  • Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
  • Labeled with six of the most commonly used solvent names or label your own; custom labeling available

Use LYOB only with solvents compatible with LDPE.

Specifications

Chemical Identifiers

500 mL
Translucent
OSHA 29 CFR 1910.1200
Polypropylene
Not Autoclavable
16 oz.
LDPE
Wide
53 mm

Specifications

500 mL
Translucent
Natural
Wide
Polypropylene
Not Autoclavable
16 oz.
LDPE
OSHA 29 CFR 1910.1200
Unlabeled
53 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions