missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Solvent-Vented™ and Safety-Labeled Wash Bottles

Pre-printed with Chemical name and formula. Fisherbrand™ Solvent-Vented™ and Safety-Labeled Wash Bottles have a patented valve which eliminates drips and allows bottle to dispense upright or inverted.
Supplier: Fisherbrand™ FB0340933
Description
- Wide mouth 2 in. (53mm) bottle for easy refilling
- Polypropylene closure with leakproof design, color-coded to aid in easy identification
- Translucent, LDPE solvent-resistant; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
- Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
- Labeled with six of the most commonly used solvent names or label your own; custom labeling available
Use LYOB only with solvents compatible with LDPE.
Chemical Identifiers
| 500 mL | |
| Translucent | |
| OSHA 29 CFR 1910.1200 | |
| Polypropylene | |
| Not Autoclavable |
| 16 oz. | |
| LDPE | |
| Wide | |
| 53 mm |
Specifications
| 500 mL | |
| Translucent | |
| Green | |
| Wide | |
| Polypropylene | |
| Not Autoclavable |
| 16 oz. | |
| LDPE | |
| OSHA 29 CFR 1910.1200 | |
| Methanol | |
| 53 mm |