missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Solvent Red 197 98.0+%, TCI America™
Spécifications
Spécifications
| Formula Weight | 381.44 |
| Percent Purity | ≥98.0% (HPLC,N) |
| Quantity | 5 g |
| Physical Form | Crystal/Powder |
| Chemical Name or Material | Solvent Red 197 |
Sécurité et manipulation
| EINECSNumber | (5)-5365 |
| TSCA | Yes |