Learn More
Eisco™ Solid Stainless Steel Hooked Weights
Solid stainless steel hooked weights with integrated hooks. Weights are cylindrical with natural stainless steel color, weight stamped at top
Supplier:  Eisco™ PHHWSS5
 Afficher plus de versions de ce produit
Numéro de catalogue. S41606
Il en reste null
Description
Description
  • Solid stainless steel construction
  • Hook is integrated with weight
  • Natural stainless steel color with stamped identifier at top
  • Weight is cylindrical
Warranty
1 year limited manufacturer warranty
Spécifications
Identifiants chimiques
Cylindrical Weight
Calibrating scales and balances
1
Stainless Steel
Weight with hook
Spécifications
Cylindrical Weight
200 g
Calibrating scales and balances
1
Stainless Steel
3.2 cm
Weight with hook
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS