Learn More
Synthware™ Solid Phase Peptide Synthesis Vessel, Ground Glass Joint, T-bore PTFE Plug
Solid Phase Peptide Synthesis Vessel with fritted glass disc and a lower ground glass joint.
$491.18 - $705.25
Specifications
| For Use With (Application) | Organic Synthesis |
|---|---|
| Material | Borosilicate Glass |
| Catalog Number | Mfr. No. | Capacity (Metric) | Product Type | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Product Type | Price | Quantity & Availability | ||||
31501589
![]() |
Synthware™
P150025M |
25 mL | Porosity: Medium |
Each for $491.18
|
|
||||
31501590
![]() |
Synthware™
P150050C |
50 mL | Porosity: Coarse |
Each for $535.43
|
|
||||
31501591
![]() |
Synthware™
P150050M |
50 mL | Porosity: Medium |
Each for $535.43
|
|
||||
31501592
![]() |
Synthware™
P150100M |
100 mL | Porosity: Medium |
Each for $628.71
|
|
||||
31501593
![]() |
Synthware™
P150250M |
250 mL | Porosity: Medium |
Each for $656.32
|
|
||||
31501594
![]() |
Synthware™
P150500M |
500 mL | Porosity: Medium |
Each for $705.25
|
|
||||
Specifications
| Organic Synthesis | |
| Borosilicate Glass |