Learn More
Synthware™ Solid Phase Peptide Synthesis Vessel, Ground Glass Joint, T-bore PTFE Plug
GREENER_CHOICE
Solid Phase Peptide Synthesis Vessel with fritted glass disc and a lower ground glass joint.
Supplier:  Synthware™ P150100M
 View more versions of this product
Catalog No. 31501592
Only null left
Description
Description
Specifications
Chemical Identifiers
Organic Synthesis
Borosilicate Glass
Specifications
Porosity: Medium
Organic Synthesis
100 mL
Borosilicate Glass
Product Suggestions
Product Suggestions