missing translation for 'onlineSavingsMsg'
Learn More

Synthware™ Solid Phase Peptide Synthesis Vessel, Ground Glass Joint, T-bore PTFE Plug
GREENER_CHOICE

Catalog No. 31501589
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
Product Type:
Porosity: Coarse
Porosity: Medium
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Product Type
31501589 25 mL Porosity: Medium
31501590 50 mL Porosity: Coarse
31501591 50 mL Porosity: Medium
31501592 100 mL Porosity: Medium
31501593 250 mL Porosity: Medium
31501594 500 mL Porosity: Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 31501589 Supplier Synthware™ Supplier No. P150025M

Solid Phase Peptide Synthesis Vessel with fritted glass disc and a lower ground glass joint.

Specifications

Product Type Porosity: Medium
Material Borosilicate Glass
Capacity (Metric) 25 mL
For Use With (Application) Organic Synthesis