Learn More
Thermo Scientific™ SOLA™ SPE Plates
Numéro de catalogue. p-4524364
Change view
Click to view available options
Description:
10 mg/2 mL
Sorbent:
HRP (Reversed Phase Polymeric)
Mixed Mode Strong Anion Exchanger, Polymeric
Mixed Mode Strong Cation Exchanger, Polymeric
Mixed Mode Weak Anion Exchanger, Polymeric
Mixed Mode Weak Cation Exchanger, Polymeric
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Description Sorbent
03150394 10 mg/2 mL HRP (Reversed Phase Polymeric)
60309002 10 mg/2 mL Mixed Mode Strong Cation Exchanger, Polymeric
60309003 10 mg/2 mL Mixed Mode Strong Anion Exchanger, Polymeric
03150402 10 mg/2 mL Mixed Mode Weak Cation Exchanger, Polymeric
03150403 10 mg/2 mL Mixed Mode Weak Anion Exchanger, Polymeric
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Numéro de catalogue. 03150394 Fournisseur Thermo Scientific™ Code fournisseur 60309001
Il en reste null
Innovative Thermo Scientific™ SOLA™ Solid Phase Extraction Plates deliver clean, highly reproducible sample extracts with lower elution volumes—for greater sensitivity, reliability and cost savings.
Obtain clean, highly reproducible sample extracts with lower elution volumes using Thermo Scientific™ SOLA™ Solid Phase Extraction (SPE) Plates. The enhanced performance gives you greater confidence in your analytical results and lowers operating costs—without compromising ease of use or requiring complex method development. Thanks to an innovative and unique frit-less design, SOLA SPE Plates deliver unparalleled performance, including:
  • Higher levels of reproducibility
  • Lower elution volumes
  • High extract cleanliness
SOLA Solid Phase Extraction Plates achieve excellent recovery levels even with low volumes of extract solvents, resulting in a more concentrated analyte and increased sensitivity. You’ll also realize additional cost and time saving from faster sample dry-down time and reduced solvent usage.
10mg/2mL 96 well plate format
Hydrophobic Reversed Phase (HRP), Strong Cation Exchanger (SCX) and Strong Anion Exchanger (SAX) phase types
SOLA products are also available in cartridge format.
Spécifications
Description 10 mg/2 mL
Bed Weight 10 mg
For Use With (Application) Bioanalytical Applications - Hydrophobic Retention of Neutral Compounds with Complementary Retention of Polar Analytes. An All-Purpose Phase.
Product Line SOLA
Sorbent HRP (Reversed Phase Polymeric)
Type 96-well SPE Plate
Volume (Metric) 2 mL
Volume (Metric) Well 2 mL
Quantity 1 Pk.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.