missing translation for 'onlineSavingsMsg'
Learn More

Sodium Vinylsulfonate (25% in Water, ca. 2.3mol/L), TCI America™

Catalog No. V0043500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V0043500ML 500 mL
V004325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V0043500ML Supplier TCI America Supplier No. V0043500ML

Chemical Identifiers

CAS 3039-83-6
Molecular Formula C2H3NaO3S
Molecular Weight (g/mol) 130.09
MDL Number MFCD00007520
InChI Key BWYYYTVSBPRQCN-UHFFFAOYSA-M
Synonym sodium ethenesulfonate, sodium vinylsulfonate, ethenesulfonic acid, sodium salt, sodium ethylenesulfonate, sodium vinyl sulfonate, sodium ethylenesulphonate, sodium apolate, unii-f7k3l38z7b, sodium ethenesulfonate in h2o, lyapolate sodium usan
PubChem CID 3270262
IUPAC Name sodium ethenesulfonate
SMILES [Na+].[O-]S(=O)(=O)C=C

Specifications

Melting Point -20°C
Quantity 500 mL
Formula Weight 130.09
Physical Form Liquid
Chemical Name or Material Sodium Vinylsulfonate (25% in Water, ca. 2.3mol/L)
EINECSNumber (2)-1588
TSCA Yes