missing translation for 'onlineSavingsMsg'
Learn More

Sodium tungsten oxide dihydrate, ACS, 99.0-101.0%

Catalog No. AA3648936
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3648936 500 g
AA3648922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3648936 Supplier Thermo Scientific Chemicals Supplier No. 03648936
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10213-10-2 | H4Na2O6W | 329.846 g/mol

In organic chemistry, sodium tungstate is used as catalyst for epoxidation of alkenes and oxidation of alcohols into aldehydes or ketones. Used in examining the serum protein, serum protein-free filtrate. Used as alkaloid precipitating agent. Used in manufacture of metal tungsten, tungstate and tungsten oxide. Waterproof and fireproof fabric.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10213-10-2
Molecular Formula H4Na2O6W
Molecular Weight (g/mol) 329.846
MDL Number MFCD00149190
InChI Key WPZFLQRLSGVIAA-UHFFFAOYSA-N
Synonym sodium tungstate dihydrate, sodium tungstate, dihydrate, unii-x1f4ny6u13, tungstic acid, sodium salt, dihydrate, tungstic acid sodium salt dihydrate, tungstic acid sodium salt, disodium dihydrate tungstate, sodium tungstate vi dihydrate, na2o4w.2h2o, na2wo4.2h2o
PubChem CID 150191
ChEBI CHEBI:63939
IUPAC Name disodium;dioxido(dioxo)tungsten;dihydrate
SMILES O.O.[O-][W](=O)(=O)[O-].[Na+].[Na+]

Specifications

Melting Point 100°C (in -2H2O)
Color White
Physical Form Powder
Quantity 500 g
Odor Odorless
Density 3.250 g/mL
Merck Index 14,8698
Solubility Information Soluble in water. Insoluble in alcohol.
Formula Weight 329.86 (293.83 Anhydrous)
Percent Purity 99.0 to 101.0%
Grade ACS
Chemical Name or Material Sodium tungsten oxide dihydrate
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
EINECSNumber 236-743-4
RTECSNumber YO7900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only