missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tripolyphosphate, Anhydrous, FCC, 85%, Spectrum™ Chemical

Catalog No. 18606900
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606900 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18606900 Supplier Spectrum Chemical Mfg Cor Supplier No. S1508500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

S1508, 7758-29-4, Na5P3O10

Spectrum™ Chemical Sodium Tripolyphosphate, Anhydrous, FCC is a food preservative and as an emulsifier to retain moisture. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7758-29-4
Molecular Formula Na5O10P3
Molecular Weight (g/mol) 367.86
InChI Key HWGNBUXHKFFFIH-UHFFFAOYSA-I
IUPAC Name pentasodium bis(phosphonatooxy)phosphinate
SMILES [Na+].[Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])(=O)OP([O-])([O-])=O

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 500 g
Percent Purity 85%
Grade FCC
Packaging Poly Bottle
Chemical Name or Material Sodium Tripolyphosphate, Anhydrous