missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate Pentahydrate Solution (0.1N (N/10)/Certified), Fisher Chemical™

Catalog No. SS36820
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Poly Bottle
PolyPac™
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SS36820 20 L PolyPac™
SS3681 1 L Poly Bottle
SS3684 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SS36820 Supplier Thermo Fisher Scientific Supplier No. FLSS36820
Edge

Standardized at 25°C against NIST™ potassium dichromate

Chemical Identifiers

CAS 10102-17-7 , 7732-18-5
Molecular Formula H10Na2O8S2
Molecular Weight (g/mol) 248.172
MDL Number MFCD00003499
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
PubChem CID 61475
ChEBI CHEBI:32150
IUPAC Name disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
SMILES O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]

Specifications

Color Colorless
Boiling Point 100°C
Melting Point 0°C
Concentration 0.0995 to 0.1005N
Grade Certified
Identification Pass Test
Packaging PolyPac™
Traceability to NIST LOT NUMBER USED POTASSIUM DICHROMATE
Vapor Pressure 14mmHg
Quantity 20 L
Physical Form Liquid
Chemical Name or Material Sodium Thiosulfate Pentahydrate Solution, 0.1N (N/10)
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category Signal Word: CAUTION!
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause eye, skin, and respiratory tract irritation . The toxicological properties have not been fully investigated.
NFPA
Health:1 Flammability:0 Instability:0