missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate, 2.00 N, Ricca Chemical

Catalog No. p-3252440
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
797516 500 mL Natural Poly Bottle
797532 1 L Natural Poly Bottle
79751 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 797516 Supplier Ricca Chemical Company Supplier No. 797516
Only null left

Chemical Identifiers

CAS 10102-17-7 , 7732-18-5 , 497-19-8
Molecular Formula H10Na2O8S2
Molecular Weight (g/mol) 248.17
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
PubChem CID 61475
ChEBI CHEBI:32150
IUPAC Name disodium pentahydrate sulfanidesulfonate
SMILES O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O

Specifications

Color Colorless
CAS Min % 32.59, 65.0, 0.26
CAS Max % 33.92, 68.0, 0.27
Grade Laboratory
Packaging Natural Poly Bottle
pH ∼10
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 2.00Normal
Chemical Name or Material Sodium Thiosulfate