missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiosulfate, 2.00 N, Ricca Chemical
Specifications
Specifications
| Color | Colorless |
| CAS Min % | 32.59, 65.0, 0.26 |
| CAS Max % | 33.92, 68.0, 0.27 |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| pH | ∼10 |
| Quantity | 500 mL |
| Physical Form | Liquid |
| Concentration or Composition Notes | 2.00Normal |
| Chemical Name or Material | Sodium Thiosulfate |