missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiosulfate, 10% (w/v), Ricca Chemical
Supplier: Ricca Chemical Company 7867632
Specifications
| Sodium Thiosulfate | |
| 10102-17-7,7732-18-5 | |
| 93.0,9.28 | |
| sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate | |
| O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+] | |
| 248.172 | |
| CHEBI:32150 | |
| Laboratory | |
| Natural Poly Bottle | |
| Liquid |
| Colorless | |
| 89.0,8.91 | |
| H10Na2O8S2 | |
| PODWXQQNRWNDGD-UHFFFAOYSA-L | |
| disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate | |
| 61475 | |
| 10% | |
| 10% (w/v) Aqueous Solution | |
| 1 L |
Chemical Identifiers
| 10102-17-7 | |
| 248.172 | |
| sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate | |
| CHEBI:32150 | |
| O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+] |
| H10Na2O8S2 | |
| PODWXQQNRWNDGD-UHFFFAOYSA-L | |
| 61475 | |
| disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate |
Safety and Handling
ShelfLife : 18 months