missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiosulfate, 0.200 N (N/5), Ricca Chemical
Supplier: Ricca Chemical Company 796025
Specifications
| Sodium Thiosulfate | |
| 10102-17-7 , 7732-18-5 , 497-19-8 | |
| 3.72,98.0,0.03 | |
| sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate | |
| O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O | |
| 248.17 | |
| CHEBI:32150 | |
| Cubitainer | |
| 10 L | |
| 0.200Normal |
| Colorless | |
| 3.57,94.0,0.03 | |
| H10Na2O8S2 | |
| PODWXQQNRWNDGD-UHFFFAOYSA-L | |
| disodium pentahydrate sulfanidesulfonate | |
| 61475 | |
| Laboratory | |
| ∼10 | |
| Liquid |
Chemical Identifiers
| 10102-17-7 | |
| 248.17 | |
| sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate | |
| CHEBI:32150 | |
| O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O |
| H10Na2O8S2 | |
| PODWXQQNRWNDGD-UHFFFAOYSA-L | |
| 61475 | |
| disodium pentahydrate sulfanidesulfonate |