missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate, 0.200 N (N/5), Ricca Chemical

Supplier:  Ricca Chemical Company 796016

 View more versions of this product

Catalog No. 796016


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Thiosulfate
10102-17-7 , 7732-18-5 , 497-19-8
3.72,98.0,0.03
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O
248.17
CHEBI:32150
Natural Poly Bottle
500 mL
0.200Normal
Colorless
3.57,94.0,0.03
H10Na2O8S2
PODWXQQNRWNDGD-UHFFFAOYSA-L
disodium pentahydrate sulfanidesulfonate
61475
Laboratory
∼10
Liquid

Chemical Identifiers

10102-17-7
248.17
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
CHEBI:32150
O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O
H10Na2O8S2
PODWXQQNRWNDGD-UHFFFAOYSA-L
61475
disodium pentahydrate sulfanidesulfonate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions