missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate, 0.100 N Aqueous Solution, APHA, Spectrum™ Chemical

Catalog No. p-7166513
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613929 500 mL Amber Glass Bottle
18613930 1 L Amber Glass Bottle
18613931 4 L Amber Glass Bottle
18613932 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18613929 Supplier Spectrum Chemical Mfg Cor Supplier No. S535500ML
Only null left

Chemical Identifiers

CAS 540-72-7 , 7732-18-5 , 10102-17-7
Molecular Formula CNNaS
Molecular Weight (g/mol) 81.07
InChI Key VGTPCRGMBIAPIM-UHFFFAOYSA-M
IUPAC Name sodium cyanosulfanide
SMILES [Na+].[S-]C#N

Specifications

Quantity 500 mL
Concentration or Composition (by Analyte or Components) 0.100 N ± 0.0005 N
Packaging Amber Glass Bottle

CAUTION: For laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.