CAS RN 540-72-7
Sodium Thiocyanate (Crystalline/Certified ACS), Fisher Chemical
CAS: 540-72-7 Molecular Formula: CNNaS Molecular Weight (g/mol): 81.07 MDL Number: MFCD00011123 InChI Key: VGTPCRGMBIAPIM-UHFFFAOYSA-M Synonym: sodium thiocyanate,sodium rhodanate,sodium sulfocyanate,sodium rhodanide,thiocyanic acid, sodium salt,thiocyanate sodium,scyan,sodium isothiocyanate,nascn,haimased PubChem CID: 516871 ChEBI: CHEBI:30952 IUPAC Name: sodium cyanosulfanide SMILES: [Na+].[S-]C#N
Sodium thiocyanate, 98+%, ACS reagent, Thermo Scientific Chemicals
CAS: 540-72-7 Molecular Formula: CNNaS Molecular Weight (g/mol): 81.07 MDL Number: MFCD00011123 InChI Key: VGTPCRGMBIAPIM-UHFFFAOYSA-M Synonym: sodium thiocyanate,sodium rhodanate,sodium sulfocyanate,sodium rhodanide,thiocyanic acid, sodium salt,thiocyanate sodium,scyan,sodium isothiocyanate,nascn,haimased PubChem CID: 516871 ChEBI: CHEBI:30952 SMILES: [Na+].[S-]C#N
Sodium thiocyanate, ACS, 98.0% min
CAS: 540-72-7 Molecular Formula: CNNaS Molecular Weight (g/mol): 81.07 MDL Number: MFCD00011123 InChI Key: VGTPCRGMBIAPIM-UHFFFAOYSA-M Synonym: sodium thiocyanate,sodium rhodanate,sodium sulfocyanate,sodium rhodanide,thiocyanic acid, sodium salt,thiocyanate sodium,scyan,sodium isothiocyanate,nascn,haimased PubChem CID: 516871 ChEBI: CHEBI:30952 SMILES: [Na+].[S-]C#N
Sodium thiocyanate, 96%
CAS: 540-72-7 Molecular Formula: CNNaS Molecular Weight (g/mol): 81.07 MDL Number: MFCD00011123 InChI Key: VGTPCRGMBIAPIM-UHFFFAOYSA-M Synonym: sodium thiocyanate,sodium rhodanate,sodium sulfocyanate,sodium rhodanide,thiocyanic acid, sodium salt,thiocyanate sodium,scyan,sodium isothiocyanate,nascn,haimased PubChem CID: 516871 ChEBI: CHEBI:30952 IUPAC Name: sodium;thiocyanate SMILES: [Na+].[S-]C#N
Sodium thiocyanate, 98%, extra pure
CAS: 540-72-7 Molecular Formula: CNNaS Molecular Weight (g/mol): 81.07 MDL Number: MFCD00011123 InChI Key: VGTPCRGMBIAPIM-UHFFFAOYSA-M Synonym: sodium thiocyanate,sodium rhodanate,sodium sulfocyanate,sodium rhodanide,thiocyanic acid, sodium salt,thiocyanate sodium,scyan,sodium isothiocyanate,nascn,haimased PubChem CID: 516871 ChEBI: CHEBI:30952 IUPAC Name: sodium;thiocyanate SMILES: [Na+].[S-]C#N
Sodium Thiocyanate, ACS Grade, ≥ 98.0%, LabChem™
CAS: 540-72-7 Molecular Formula: CNNaS Molecular Weight (g/mol): 81.07 MDL Number: MFCD00011123 InChI Key: VGTPCRGMBIAPIM-UHFFFAOYSA-M Synonym: sodium thiocyanate,sodium rhodanate,sodium sulfocyanate,sodium rhodanide,thiocyanic acid, sodium salt,thiocyanate sodium,scyan,sodium isothiocyanate,nascn,haimased PubChem CID: 516871 ChEBI: CHEBI:30952 IUPAC Name: sodium cyanosulfanide SMILES: [Na+].[S-]C#N
Sodium Thiocyanate, ACS Reagent Grade, Ricca Chemical
CAS: 540-72-7 Molecular Formula: CNNaS Molecular Weight (g/mol): 81.07 MDL Number: MFCD00011123 InChI Key: VGTPCRGMBIAPIM-UHFFFAOYSA-M Synonym: sodium thiocyanate,sodium rhodanate,sodium sulfocyanate,sodium rhodanide,thiocyanic acid, sodium salt,thiocyanate sodium,scyan,sodium isothiocyanate,nascn,haimased PubChem CID: 516871 ChEBI: CHEBI:30952 IUPAC Name: sodium cyanosulfanide SMILES: [Na+].[S-]C#N
Sodium Thiocyanate, 0.100 N (N/10), Ricca Chemical
CAS: 540-72-7 Molecular Formula: CNNaS Molecular Weight (g/mol): 81.07 MDL Number: MFCD00011123 InChI Key: VGTPCRGMBIAPIM-UHFFFAOYSA-M Synonym: sodium thiocyanate,sodium rhodanate,sodium sulfocyanate,sodium rhodanide,thiocyanic acid, sodium salt,thiocyanate sodium,scyan,sodium isothiocyanate,nascn,haimased PubChem CID: 516871 ChEBI: CHEBI:30952 IUPAC Name: sodium cyanosulfanide SMILES: [Na+].[S-]C#N
Sodium Thiosulfate, 1.0 N Aqueous Solution, Spectrum™ Chemical
CAS: 540-72-7 Molecular Formula: CNNaS Molecular Weight (g/mol): 81.07 InChI Key: VGTPCRGMBIAPIM-UHFFFAOYSA-M IUPAC Name: sodium cyanosulfanide SMILES: [Na+].[S-]C#N
Sodium Thiocyanate, ACS Reagent, Reagents
CAS: 540-72-7 Molecular Formula: CNNaS Molecular Weight (g/mol): 81.07 InChI Key: VGTPCRGMBIAPIM-UHFFFAOYSA-M Synonym: Sodium rhodanate, Sodium rhodanide, Sodium sulfocyanate IUPAC Name: sodium cyanosulfanide SMILES: [Na+].[S-]C#N
Sodium Thiosulfate, 0.100 N Aqueous Solution, APHA, Spectrum™ Chemical
CAS: 540-72-7 Molecular Formula: CNNaS Molecular Weight (g/mol): 81.07 InChI Key: VGTPCRGMBIAPIM-UHFFFAOYSA-M IUPAC Name: sodium cyanosulfanide SMILES: [Na+].[S-]C#N
Sodium Thiosulfate, 0.01 N Aqueous Solution, Spectrum™ Chemical
CAS: 540-72-7 Molecular Formula: CNNaS Molecular Weight (g/mol): 81.07 InChI Key: VGTPCRGMBIAPIM-UHFFFAOYSA-M IUPAC Name: sodium cyanosulfanide SMILES: [Na+].[S-]C#N