missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate, 0.0500 N (N/20), Ricca Chemical

Catalog No. 794032
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
794032 1 L Natural Poly Bottle
79401 4 L Natural Poly Bottle
79405 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 794032 Supplier Ricca Chemical Company Supplier No. 794032

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10102-17-7 , 7732-18-5 , 497-19-8
Molecular Formula H10Na2O8S2
Molecular Weight (g/mol) 248.17
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
PubChem CID 61475
ChEBI CHEBI:32150
IUPAC Name disodium pentahydrate sulfanidesulfonate
SMILES O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O

Specifications

Color Colorless
CAS Min % 0.97, 97.0, 0.01
CAS Max % 1.01, 100.0, 0.01
Grade Laboratory
Packaging Natural Poly Bottle
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 0.0500Normal
Chemical Name or Material Sodium Thiosulfate