missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiosulfate, 0.0250 N (N/40), Ricca Chemical
Specifications
Specifications
| CAS Min % | 0.49, 97.0, 0.24, 0.0039 |
| CAS Max % | 0.51, 100.0, 0.24, 0.0041 |
| SMILES | O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O |
| IUPAC Name | disodium pentahydrate sulfanidesulfonate |
| Molecular Weight (g/mol) | 248.17 |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| Quantity | 500 mL |
| Concentration or Composition Notes | 0.0250N |
| Chemical Name or Material | Sodium Thiosulfate |