missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate, 0.0250 N (N/40), Ricca Chemical

Catalog No. 79005
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
79005 20 L Cubitainer
790016 500 mL Natural Poly Bottle
790032 1 L Natural Poly Bottle
R79000004F 4 L Cubitainer
79001 4 L Natural Poly Bottle
790025 10 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 79005 Supplier Ricca Chemical Company Supplier No. 79005
Only null left

Chemical Identifiers

CAS 10102-17-7 , 7732-18-5 , 71-41-0 , 497-19-8
Molecular Formula H10Na2O8S2
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
PubChem CID 61475
ChEBI CHEBI:32150

Specifications

CAS Min % 0.49, 97.0, 0.24, 0.0039
CAS Max % 0.51, 100.0, 0.24, 0.0041
SMILES O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O
IUPAC Name disodium pentahydrate sulfanidesulfonate
Molecular Weight (g/mol) 248.17
Grade Laboratory
Packaging Cubitainer
Quantity 20 L
Concentration or Composition Notes 0.0250N
Chemical Name or Material Sodium Thiosulfate