missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiosulfate, 0.0250 N (N/40), Ricca Chemical
Supplier: Ricca Chemical Company 790032
Specifications
| Sodium Thiosulfate | |
| 0.49,97.0,0.24,0.0039 | |
| H10Na2O8S2 | |
| PODWXQQNRWNDGD-UHFFFAOYSA-L | |
| disodium pentahydrate sulfanidesulfonate | |
| 61475 | |
| Laboratory | |
| 1 L |
| 10102-17-7,7732-18-5,71-41-0,497-19-8 | |
| 0.51,100.0,0.24,0.0041 | |
| sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate | |
| O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O | |
| 248.17 | |
| CHEBI:32150 | |
| Natural Poly Bottle | |
| 0.0250N |
Chemical Identifiers
| 10102-17-7 | |
| PODWXQQNRWNDGD-UHFFFAOYSA-L | |
| 61475 |
| H10Na2O8S2 | |
| sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate | |
| CHEBI:32150 |