missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate, 0.0250 N (N/40), Ricca Chemical

Supplier:  Ricca Chemical Company 79001

 View more versions of this product

Catalog No. 7900-1

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Thiosulfate
0.49,97.0,0.24,0.0039
H10Na2O8S2
PODWXQQNRWNDGD-UHFFFAOYSA-L
disodium pentahydrate sulfanidesulfonate
61475
Laboratory
4 L
10102-17-7,7732-18-5,71-41-0,497-19-8
0.51,100.0,0.24,0.0041
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O
248.17
CHEBI:32150
Natural Poly Bottle
0.0250N

Chemical Identifiers

10102-17-7
PODWXQQNRWNDGD-UHFFFAOYSA-L
61475
H10Na2O8S2
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
CHEBI:32150
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions