missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate, 0.0100 N (N/100), Ricca Chemical

Supplier:  Ricca Chemical Company R78900004F

 View more versions of this product

Catalog No. R78900004F

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

SODIUM THIOSULFATE
0.2,98.0,0.0016
H10Na2O8S2
PODWXQQNRWNDGD-UHFFFAOYSA-L
disodium pentahydrate sulfanidesulfonate
61475
Laboratory
4 L
10102-17-7,7732-18-5,497-19-8
0.21,100.0,0.0016
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O
248.17
CHEBI:32150
Cubitainer

Chemical Identifiers

10102-17-7
248.17
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
CHEBI:32150
O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O
H10Na2O8S2
PODWXQQNRWNDGD-UHFFFAOYSA-L
61475
disodium pentahydrate sulfanidesulfonate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions