missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate, 0.0100 N (N/100), Ricca Chemical

Catalog No. p-2421826
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
789016 500 mL Natural Poly Bottle
789032 1 L Natural Poly Bottle
R78900004F 4 L Cubitainer
78901 4 L Natural Poly Bottle
78905 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 789016 Supplier Ricca Chemical Company Supplier No. 789016
Only null left

Chemical Identifiers

CAS 10102-17-7 , 7732-18-5 , 497-19-8
Molecular Formula H10Na2O8S2
Molecular Weight (g/mol) 248.17
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
PubChem CID 61475
ChEBI CHEBI:32150
IUPAC Name disodium pentahydrate sulfanidesulfonate
SMILES O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O

Specifications

CAS Min % 0.2, 98.0, 0.0016
CAS Max % 0.21, 100.0, 0.0016
Grade Laboratory
Packaging Natural Poly Bottle
Quantity 500 mL
Chemical Name or Material SODIUM THIOSULFATE