missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiosulfate, 0.0100 N (N/100), Ricca Chemical
Specifications
Specifications
| CAS Min % | 0.2, 98.0, 0.0016 |
| CAS Max % | 0.21, 100.0, 0.0016 |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| Quantity | 500 mL |
| Chemical Name or Material | SODIUM THIOSULFATE |