missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate, 0.01 N Aqueous Solution, Spectrum™ Chemical

Catalog No. p-7166510
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
Catalog No. Quantity Packaging
18613915 500 mL Amber Glass Bottle
18613916 1 L Amber Glass Bottle
18613917 4 L Amber Glass Bottle
18613918 20 L Cubitainer
4 options

Catalog No. 18613915

Supplier: Spectrum Chemical Mfg Cor S527500ML

Only null left

Chemical Identifiers

CAS 540-72-7 , 7732-18-5 , 10102-17-7
Molecular Formula CNNaS
Molecular Weight (g/mol) 81.07
InChI Key VGTPCRGMBIAPIM-UHFFFAOYSA-M
IUPAC Name sodium cyanosulfanide
SMILES [Na+].[S-]C#N

Specifications

Quantity 500 mL
Concentration or Composition (by Analyte or Components) 0.01 N ± 0.0002 N
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.