missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiomethoxide, 95%, pure

Numéro de catalogue. AC305710050
Modifier l'affichage
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
AC305710050 5 g Glass bottle
AC305710010 1 g Glass bottle
AC305710250 25 g Glass bottle
AC305711000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. AC305710050 Fournisseur Thermo Scientific Chemicals Code fournisseur 305710050
Il en reste null

CAS: 5188-07-8 | CH3NaS | 70.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 5188-07-8
Molecular Formula CH3NaS
Molecular Weight (g/mol) 70.09
MDL Number MFCD00174316
InChI Key RMBAVIFYHOYIFM-UHFFFAOYSA-M
PubChem CID 4378561
SMILES C[S-].[Na+]

Spécifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 94 to 106% (Iodimetry)
Packaging Glass bottle
Linear Formula CH3SNa
Quantity 5 g
Synonym sodium thiomethoxide, sodium methanethiolate, sodium thiomethylate, methyl mercaptan sodium salt, methanethiol sodium salt, sodiumthiomethoxide, methanethiol, sodium salt, sodiummethanethiolate, sodium methyl mercaptide, methylsulfanyl sodium
Solubility Information Solubility in water: freely soluble.
IUPAC Name sodium;methanethiolate
Formula Weight 70.09
Percent Purity 95%
Grade Pure
Physical Form Powder
Chemical Name or Material Sodium thiomethoxide
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable solid.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
EINECSNumber 225-969-9

RUO – Research Use Only