missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiomethoxide, 90+%, technical

Catalog No. AC469891000
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469891000 100 g
AC469890010 1 g
AC469890050 5 g
AC469890250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC469891000 Supplier Thermo Scientific Chemicals Supplier No. 469891000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5188-07-8 | CH3NaS | 70.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5188-07-8
Molecular Formula CH3NaS
Molecular Weight (g/mol) 70.08
InChI Key RMBAVIFYHOYIFM-UHFFFAOYSA-M
IUPAC Name sodium methylsulfanide
SMILES [Na+].C[S-]

Specifications

Assay Percent Range 90+%
Linear Formula CH3SNa
Quantity 100 g
Solubility Information Solubility aq. soln.: freely soluble.
Formula Weight 70.09
Grade Technical
Chemical Name or Material Sodium thiomethoxide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Flammable solid.

Toxic if swallowed.

Causes severe skin burns and eye damage.

Precautionary Statement GHS P Statement Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.

No smoking.

Wear protective gloves/protective clothing/eye protection/face protection.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Immediately call a POISON CENTER or doctor/physician.

IF SWALLOWED: Rinse mouth.

Do NOT induce vomiting.

IF ON SKIN (or hair): Take off immediately all contaminated clothing.

Rinse skin with water or shower
EINECSNumber 225-969-9

RUO – Research Use Only