missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiocyanate, ACS Reagent Grade, Ricca Chemical

Catalog No. RDCS0910500
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCS0910500 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCS0910500 Supplier Ricca Chemical Company Supplier No. RDCS0910500B1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 540-72-7
Molecular Formula CNNaS
Molecular Weight (g/mol) 81.07
MDL Number MFCD00011123
InChI Key VGTPCRGMBIAPIM-UHFFFAOYSA-M
Synonym sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased
PubChem CID 516871
ChEBI CHEBI:30952
IUPAC Name sodium cyanosulfanide
SMILES [Na+].[S-]C#N

Specifications

CAS Min % 98.0
CAS Max % 100.0
Physical Form Fine Crystals or Crystalline Powder
Quantity 500 g
Percent Purity ≥ 98%
Grade ACS
Packaging White Poly Wide Mouth Bottle
Chemical Name or Material Sodium Thiocyanate
ShelfLife 60 months