missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiocyanate, ACS Grade, ≥ 98.0%, LabChem™

Catalog No. LC249601
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC249601 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC249601 Supplier LabChem Supplier No. LC249601

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 540-72-7
Molecular Formula CNNaS
Molecular Weight (g/mol) 81.07
MDL Number MFCD00011123
InChI Key VGTPCRGMBIAPIM-UHFFFAOYSA-M
Synonym sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased
PubChem CID 516871
ChEBI CHEBI:30952
IUPAC Name sodium cyanosulfanide
SMILES [Na+].[S-]C#N

Specifications

Melting Point 287°C
Color White
CAS Max % 100
Physical Form Crystalline
Quantity 500 g
Linear Formula NaSCN
Solubility Information Soluble in water
Formula Weight 81.07
Percent Purity ≥98.0%
Grade ACS
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1.74g/cm3
Decomposition Information Hydrogen cyanide; Carbon monoxide; Carbon dioxide; Sulfur compounds
Density 1.74g/cm3
Chemical Name or Material Sodium Thiocyanate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Harmful if swallowed, in contact with skin or if inhaled.
Precautionary Statement GHS P Statement
Avoid breathing dust.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin: Wash with plenty of soap and water.
Call a poison center/doctor if you feel unwell.
Take off contaminated clothing and wash it before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
Dispose of contents/container to comply with local, state and federal regulations.
EINECSNumber 208-754-4
Recommended Storage Room Temperature