missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiocyanate, 0.100 N (N/10), Ricca Chemical

Catalog No. 786016
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
786016 500 mL Natural Poly Bottle
786032 1 L Natural Poly Bottle
78601 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 786016 Supplier Ricca Chemical Company Supplier No. RI786016

Chemical Identifiers

CAS 540-72-7 , 7732-18-5
Molecular Formula CNNaS
Molecular Weight (g/mol) 81.07
MDL Number MFCD00011123
InChI Key VGTPCRGMBIAPIM-UHFFFAOYSA-M
Synonym sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased
PubChem CID 516871
ChEBI CHEBI:30952
IUPAC Name sodium cyanosulfanide
SMILES [Na+].[S-]C#N

Specifications

Color Colorless
CAS Min % 97.0, 0.79
CAS Max % 100.0, 0.82
Grade Laboratory
Packaging Natural Poly Bottle
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 0.100Normal
Chemical Name or Material Sodium Thiocyanate