missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiocyanate, 0.100 N (N/10), Ricca Chemical

Supplier:  Ricca Chemical Company 78601

 View more versions of this product

Catalog No. 7860-1


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Thiocyanate
540-72-7 , 7732-18-5
100.0,0.82
MFCD00011123
VGTPCRGMBIAPIM-UHFFFAOYSA-M
sodium cyanosulfanide
516871
Laboratory
4 L
0.100Normal
Colorless
97.0,0.79
CNNaS
sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased
[Na+].[S-]C#N
81.07
CHEBI:30952
Natural Poly Bottle
Liquid

Chemical Identifiers

540-72-7
81.07
VGTPCRGMBIAPIM-UHFFFAOYSA-M
516871
sodium cyanosulfanide
CNNaS
MFCD00011123
sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased
CHEBI:30952
[Na+].[S-]C#N
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions