missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tetrakis(4-fluorophenyl)borate Hydrate 98.0+%, TCI America™

Catalog No. A51315G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A51315G 5 g
A51311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A51315G Supplier TCI America Supplier No. A51315G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

[Precipitation reagent for Cs and titrimetric reagent for nonionic surfactants]

Chemical Identifiers

CAS 25776-12-9
Molecular Formula C24H20BF4NaO2
Molecular Weight (g/mol) 450.215
MDL Number MFCD00010732
InChI Key MSDGDEJOIBMWJD-UHFFFAOYSA-N
Synonym Tetrakis(4-fluorophenyl)boron Sodium
PubChem CID 45073643
IUPAC Name sodium;tetrakis(4-fluorophenyl)boranuide;dihydrate
SMILES [B-](C1=CC=C(C=C1)F)(C2=CC=C(C=C2)F)(C3=CC=C(C=C3)F)C4=CC=C(C=C4)F.O.O.[Na+]

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 414.19
Percent Purity ≥98.0% (HPLC,W)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Tetrakis(4-fluorophenyl)borate Hydrate [Precipitation reagent for Cs and titrimetric reagent
TSCA No
Recommended Storage Refrigerator