missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium tetraborate decahydrate, 99.5+%, ACS reagent
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 75.0°C |
| Color | White |
| pH | 9.15 to 9.20 (0.01M soln. at 25°C) |
| Physical Form | Crystalline Powder |
| Quantity | 25 g |
| Packaging | Plastic bottle |
| Assay Percent Range | 99.5+% |
| Linear Formula | Na2B4O7·10H2O |
| Fieser | 04,461 |
| Merck Index | 15, 8726 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May damage fertility. May damage the unborn child. |
| Precautionary Statement | GHS P Statement Obtain special instructions before use. IF exposed or concerned: Get medical advice/attention. |
| RTECSNumber | VZ2275000 |
| TSCA | TSCA |
RUO – Research Use Only