missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tartrate, Dihydrate, Crystal, Reagent, ACS, 99-101%, Spectrum™ Chemical

Catalog No. 18606868
Change view
Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
25 kg
4 x 2.5 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606868 500 g Poly Bottle
18606870 2.5 kg Poly Bottle
18606869 6 x 500 g Poly Bottle
18606871 4 x 2.5 kg Poly Bottle
18606872 12 kg Poly Pail
18606873 25 kg Fiber Drum
18606874 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18606868 Supplier Spectrum Chemical Mfg Cor Supplier No. S1485500GM

S1485, 6106-24-7, Na2C4H4O6·2H2O

Spectrum™ Chemical Sodium Tartrate, Dihydrate, Crystal, Reagent, ACS has a crystal structure that is able to capture a very precise amount of water and is used in the Karl Fischer moisture determination titration technique. As an ACS grade reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 6106-24-7
Molecular Formula C4H8Na2O8
Molecular Weight (g/mol) 230.08
MDL Number MFCD00150035
InChI Key FGJLAJMGHXGFDE-UDMGOBQRNA-L
IUPAC Name disodium (2R,3R)-2,3-dihydroxybutanedioate dihydrate
SMILES O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

Specifications

CAS Max % 1
pH 7.0 to 9.0
Loss on Drying 15.61-15.71%
Packaging Poly Bottle
Quantity 500 g
Percent Purity 99.0 to 101%
Grade ACS